Ethyl [(4-iodophenyl)amino]acetate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 2-(4-iodoanilino)acetate
OR8095-1G
OR8095-5G
OR8095-10G
APOSOR8095-1GEA
116
EUR
APOSOR8095-1G
APOSOR8095-5G
APOSOR8095-10G
Ethyl [(4-iodophenyl)amino]acetate 97%
Ethyl [(4-iodophenyl)amino]acetate
Formula:
C₁₀H₁₂INO₂ MW: 305.112 g/mol Melting Pt: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00176764 CAS Number: 14108-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...