2-Chloro-4-morpholinopyridine 97%

Supplier: Apollo Scientific

OR901747-1G OR901747-5G
APOSOR901747-1GEA 255 EUR
APOSOR901747-1G APOSOR901747-5G
2-Chloro-4-morpholinopyridine 97%
2-Chloro-4-morpholinopyridine
Formula: C₉H₁₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD12923071
CAS Number: 937202-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR