Ethyl 1-isocyanocyclohexanecarboxylate 95%

Supplier: Apollo Scientific

OR905924-1G OR905924-250MG
APOSOR905924-1GEA 500 EUR
APOSOR905924-1G APOSOR905924-250MG
Ethyl 1-isocyanocyclohexanecarboxylate 95%
Ethyl 1-isocyanocyclohexanecarboxylate
Formula: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD02664648
CAS Number: 97846-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR