2-Methyleneglutaric acid 96%
Supplier: Apollo Scientific
Warning
OR908110-1G OR908110-250MG
APOSOR908110-1GEA 604 EUR
APOSOR908110-1G APOSOR908110-250MG
2-Methyleneglutaric acid 96%
2-Methyleneglutaric acid
Formula: C₆H₈O₄
Melting Pt: 131…132 °C
Storage Temperature: Freezer
MDL Number: MFCD06208028
CAS Number: 3621-79-2