4-(5-Bromothiazol-2-yl)morpholine 98%
Supplier: Apollo Scientific
Danger
OR908995-10G OR908995-1G OR908995-5G
APOSOR908995-10GEA 752 EUR
APOSOR908995-10G APOSOR908995-1G APOSOR908995-5G
4-(5-Bromothiazol-2-yl)morpholine 98%
4-(5-Bromothiazol-2-yl)morpholine
Formula: C₇H₉BrN₂OS
MW: 249.128 g/mol
Melting Pt: 90…110 °C
Storage Temperature: Ambient
MDL Number: MFCD09879069
CAS Number: 933728-73-5
UN: 2811
ADR: 6.1 III