N-(3-Cyanophenyl)-2,5-dimethylpyrrole 98%
Supplier: Apollo Scientific
OR909573-1G OR909573-25G OR909573-5G
APOSOR909573-1GEA 145 EUR
APOSOR909573-1G APOSOR909573-25G APOSOR909573-5G
N-(3-Cyanophenyl)-2,5-dimethylpyrrole 98%
N-(3-Cyanophenyl)-2,5-dimethylpyrrole
Formula: C₁₃H₁₂N₂
MW: 196.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09063237
CAS Number: 204142-42-7