2-Ethanesulphony)nicotinonitrile 98%

Supplier: Apollo Scientific

OR911102-10G OR911102-1G OR911102-5G
APOSOR911102-10GEA 754 EUR
APOSOR911102-10G APOSOR911102-1G APOSOR911102-5G
2-Ethanesulphony)nicotinonitrile 98%
2-Ethanesulphony)nicotinonitrile
Formula: C₈H₈N₂O₂S
MW: 196.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24448799
CAS Number: 1437794-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR