5-Methoxyindoline hydrochloride 95%
Supplier: Apollo Scientific
OR912195-1G OR912195-5G
APOSOR912195-1GEA 41.6 EUR
APOSOR912195-1G APOSOR912195-5G
5-Methoxyindoline hydrochloride 95%
5-Methoxyindoline hydrochloride
Formula: C₉H₁₁NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD07783777
CAS Number: 4770-39-2