1,2,3,4-Tetrahydro-2-naphthoic acid 96%

Supplier: Apollo Scientific

OR916846-1G OR916846-5G
APOSOR916846-1GEA 112 EUR
APOSOR916846-1G APOSOR916846-5G
1,2,3,4-Tetrahydro-2-naphthoic acid 96%
1,2,3,4-Tetrahydro-2-naphthoic acid
Formula: C₁₁H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00001741
CAS Number: 53440-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR