2-(7-Methoxy-3,4-dihydronaphthalen-1-yl)acetonitrile 95%
Supplier: Apollo Scientific
OR918258-25G OR918258-5G
APOSOR918258-25GEA 757 EUR
APOSOR918258-25G APOSOR918258-5G
2-(7-Methoxy-3,4-dihydronaphthalen-1-yl)acetonitrile 95%
2-(7-Methoxy-3,4-dihydronaphthalen-1-yl)acetonitrile
Formula: C₁₃H₁₃NO
MW: 199.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09840884
CAS Number: 861960-34-1