4-((2-Chloroethyl)(methyl)amino)benzaldehyde 98%

Supplier: Apollo Scientific

OR918636-100G OR918636-25G
APOSOR918636-100GEA 208 EUR
APOSOR918636-100G APOSOR918636-25G
4-((2-Chloroethyl)(methyl)amino)benzaldehyde 98%
4-((2-Chloroethyl)(methyl)amino)benzaldehyde
Formula: C₁₀H₁₂ClNO
MW: 197.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071777
CAS Number: 94-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR