tert-Butyl 2-hydroxycyclohexylcarbamate 96%
Supplier: Apollo Scientific
Synonyms: tert-Butyl (2-hydroxycyclohexyl)carbamate
OR921403-1G OR921403-25G OR921403-5G
APOSOR921403-1GEA 43.5 EUR
APOSOR921403-1G APOSOR921403-25G APOSOR921403-5G
tert-Butyl 2-hydroxycyclohexylcarbamate 96%
tert-Butyl 2-hydroxycyclohexylcarbamate
Formula: C₁₁H₂₁NO₃
MW: 215.289 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08061947
CAS Number: 477584-30-8