Imidazo[1,2-a]pyridin-3-ylmethanol 95%
Supplier: Apollo Scientific
OR921772-1G OR921772-250MG
APOSOR921772-1GEA 828 EUR
APOSOR921772-1G APOSOR921772-250MG
Imidazo[1,2-a]pyridin-3-ylmethanol 95%
Imidazo[1,2-a]pyridin-3-ylmethanol
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864457
CAS Number: 30489-43-1