Thiochroman-3-one 97%
Supplier: Apollo Scientific
OR922713-1G OR922713-250MG
APOSOR922713-1GEA 647 EUR
APOSOR922713-1G APOSOR922713-250MG
Thiochroman-3-one 97%
Thiochroman-3-one
Formula: C₉H₈OS
Storage Temperature: Ambient
MDL Number: MFCD08437618
CAS Number: 16895-58-2