2-Chloro-3-hydroxyquinoxaline 95%
Supplier: Apollo Scientific
OR922814-1G
OR922814-250MG
APOSOR922814-1GEA
806
EUR
APOSOR922814-1G
APOSOR922814-250MG
2-Chloro-3-hydroxyquinoxaline 95%
2-Chloro-3-hydroxyquinoxaline
Formula:
C₈H₅ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD14728011 CAS Number: 35676-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...