2-Hydroxy-1-(4-methylphenyl)ethanone 95%
Supplier: Apollo Scientific
Synonyms: 2-Hydroxy-4'-methylacetophenone
OR923270-1G OR923270-5G
APOSOR923270-1GEA 148 EUR
APOSOR923270-1G APOSOR923270-5G
2-Hydroxy-1-(4-methylphenyl)ethanone 95%
2-Hydroxy-1-(4-methylphenyl)ethanone
Formula: C₉H₁₀O₂
MW: 150.175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778990
CAS Number: 4079-54-3