2-(5-Methyl-1H-pyrazol-3-yl)pyridine 98%
Supplier: Apollo Scientific
OR924774-1G OR924774-250MG OR924774-5G
APOSOR924774-1GEA 137 EUR
APOSOR924774-1G APOSOR924774-250MG APOSOR924774-5G
2-(5-Methyl-1H-pyrazol-3-yl)pyridine 98%
2-(5-Methyl-1H-pyrazol-3-yl)pyridine
Formula: C₉H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD11840239
CAS Number: 19959-77-4