2-(5-Methyl-2-furyl)ethanamine 95%
Supplier: Apollo Scientific
OR925847-1G OR925847-250MG
APOSOR925847-1GEA 647 EUR
APOSOR925847-1G APOSOR925847-250MG
2-(5-Methyl-2-furyl)ethanamine 95%
2-(5-Methyl-2-furyl)ethanamine
Formula: C₇H₁₁NO
MW: 125.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059767
CAS Number: 30796-85-1