2-Ethoxy-6-hydroxy-nicotinonitrile 95%

Supplier: Apollo Scientific

OR926667-1G OR926667-5G
APOSOR926667-1GEA 219 EUR
APOSOR926667-1G APOSOR926667-5G
2-Ethoxy-6-hydroxy-nicotinonitrile 95%
2-Ethoxy-6-hydroxy-nicotinonitrile
Formula: C₈H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD23115409
CAS Number: 71350-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR