1-(1-Isobutylcyclobutyl)methanamine 95%
Supplier: Apollo Scientific
OR927139-1G
OR927139-250MG
APOSOR927139-1GEA
647
EUR
APOSOR927139-1G
APOSOR927139-250MG
1-(1-Isobutylcyclobutyl)methanamine 95%
1-(1-Isobutylcyclobutyl)methanamine
Formula:
C₉H₁₉N Storage Temperature: Ambient |
MDL Number:
MFCD09971229 CAS Number: 1015846-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...