7-Nitroisoquinolin-1(2H)-one 97%
Supplier: Apollo Scientific
OR927390-100MG OR927390-1G OR927390-250MG
APOSOR927390-100MGEA 0 EUR
APOSOR927390-100MG APOSOR927390-1G APOSOR927390-250MG
7-Nitroisoquinolin-1(2H)-one 97%
7-Nitroisoquinolin-1(2H)-one
Formula: C₉H₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD15526743
CAS Number: 20141-83-7