3-(hydroxymethyl)-4-isopropoxybenzaldehyde 95%

Supplier: Apollo Scientific

OR927479-1G OR927479-250MG
APOSOR927479-1GEA 290 EUR
APOSOR927479-1G APOSOR927479-250MG
3-(hydroxymethyl)-4-isopropoxybenzaldehyde 95%
3-(hydroxymethyl)-4-isopropoxybenzaldehyde
Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03180282
CAS Number: 959237-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR