2-Dimethylaminomethylene-5,5-dimethyl-cyclohexane-1,3-dione 95%
Supplier: Apollo Scientific
Synonyms: 2-((Dimethylamino)methylene)-5,5-dimethylcyclohexane-1,3-dione
OR927660-1G OR927660-5G
APOSOR927660-1GEA 333 EUR
APOSOR927660-1G APOSOR927660-5G
2-Dimethylaminomethylene-5,5-dimethyl-cyclohexane-1,3-dione 95%
2-Dimethylaminomethylene-5,5-dimethyl-cyclohexane-1,3-dione
Formula: C₁₁H₁₇NO₂
MW: 195.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00154288
CAS Number: 75039-59-7