4-Chloro-cinnoline 97%

Supplier: Apollo Scientific

OR928827-1G OR928827-250MG
APOSOR928827-1GEA 264 EUR
APOSOR928827-1G APOSOR928827-250MG
4-Chloro-cinnoline 97%
4-Chloro-cinnoline
Formula: C₈H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD09033775
CAS Number: 5152-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR