Methyl 1-bromocyclopropanecarboxylate 97%
Supplier: Apollo Scientific
OR929469-100G OR929469-25G
APOSOR929469-100GEA 102 EUR
APOSOR929469-100G APOSOR929469-25G
Methyl 1-bromocyclopropanecarboxylate 97%
Methyl 1-bromocyclopropanecarboxylate
Formula: C₅H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD12922645
CAS Number: 96999-01-8