N-Methyl-3-cyano-6-hydroxy-4-methyl-2-pyridone 95%
Supplier: Apollo Scientific
OR929912-1G OR929912-250MG
APOSOR929912-1GEA 357 EUR
APOSOR929912-1G APOSOR929912-250MG
N-Methyl-3-cyano-6-hydroxy-4-methyl-2-pyridone 95%
N-Methyl-3-cyano-6-hydroxy-4-methyl-2-pyridone
Formula: C₈H₈N₂O₂
MW: 164.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044065
CAS Number: 27074-03-9