(S)-1-Methoxy-3-phenyl-2-propylamine 90%
Supplier: Apollo Scientific
OR930429-1G OR930429-5G
APOSOR930429-1GEA 248 EUR
APOSOR930429-1G APOSOR930429-5G
(S)-1-Methoxy-3-phenyl-2-propylamine 90%
(S)-1-Methoxy-3-phenyl-2-propylamine
Formula: C₁₀H₁₅NO
MW: 165.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318301
CAS Number: 64715-80-6