Aminotriester 95%
Supplier: Apollo Scientific
Warning
Synonyms: Di-tert-butyl 4-amino-4-(3-(tert-butoxy)-3-oxopropyl)heptanedioate
OR931286-1G OR931286-5G OR931286-250MG
APOSOR931286-1GEA 180 EUR
APOSOR931286-1G APOSOR931286-5G APOSOR931286-250MG
Aminotriester 95%
Aminotriester
Formula: C₂₂H₄₁NO₆
MW: 415.564 g/mol
Melting Pt: 60…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07357283
CAS Number: 136586-99-7