4,7-Dichloro-1,6-naphthyridine 96%
Supplier: Apollo Scientific
OR932711-1G OR932711-250MG
APOSOR932711-1GEA 467 EUR
APOSOR932711-1G APOSOR932711-250MG
4,7-Dichloro-1,6-naphthyridine 96%
4,7-Dichloro-1,6-naphthyridine
Formula: C₈H₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD11044711
CAS Number: 952138-13-5