6-Chloro-2-ethyl-1H-benzimidazole 95%
Supplier: Apollo Scientific
Synonyms: 6-Chloro-2-ethyl-1H-benzo[d]imidazole
OR932774-1G OR932774-250MG OR932774-5G
APOSOR932774-1GEA 115 EUR
APOSOR932774-1G APOSOR932774-250MG APOSOR932774-5G
6-Chloro-2-ethyl-1H-benzimidazole 95%
6-Chloro-2-ethyl-1H-benzimidazole
Formula: C₉H₉ClN₂
MW: 180.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18800923
CAS Number: 34569-15-8