5-Methoxy-4-methylindole-3-carboxaldehyde 95%
Supplier: Apollo Scientific
OR938673-100MG OR938673-1G OR938673-250MG
APOSOR938673-100MGEA 145 EUR
APOSOR938673-100MG APOSOR938673-1G APOSOR938673-250MG
5-Methoxy-4-methylindole-3-carboxaldehyde 95%
5-Methoxy-4-methylindole-3-carboxaldehyde
Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265112
CAS Number: 932710-64-0