1-(2,2-Dimethoxyethyl)-4-methoxybenzene 95%
Supplier: Apollo Scientific
OR940012-1G OR940012-5G
APOSOR940012-1GEA 258 EUR
APOSOR940012-1G APOSOR940012-5G
1-(2,2-Dimethoxyethyl)-4-methoxybenzene 95%
1-(2,2-Dimethoxyethyl)-4-methoxybenzene
Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD15144799
CAS Number: 42866-92-2