1-Methyl-1H-indol-2-amine hydrochloride 95%
Supplier: Apollo Scientific
OR941089-1G OR941089-250MG
APOSOR941089-1GEA 632 EUR
APOSOR941089-1G APOSOR941089-250MG
1-Methyl-1H-indol-2-amine hydrochloride 95%
1-Methyl-1H-indol-2-amine hydrochloride
Formula: C₉H₁₀N₂·ClH
MW: 182.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143632
CAS Number: 42456-82-6