3-Methyloxolan-3-ol 95%

Supplier: Apollo Scientific
Warning

OR943342-1G OR943342-250MG
APOSOR943342-1GEA 661 EUR
APOSOR943342-1G APOSOR943342-250MG
3-Methyloxolan-3-ol 95%
3-Methyloxolan-3-ol
Formula: C₅H₁₀O₂
MW: 102.132 g/mol
Boiling Pt: 76…80 °C (18 mm Hg)
Density: 1.032 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD18332778
CAS Number: 64360-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR