3-(2-Methylpropyl)-1,2-oxazol-5-amine 95%
Supplier: Apollo Scientific
OR943865-1G OR943865-250MG
APOSOR943865-1GEA 770 EUR
APOSOR943865-1G APOSOR943865-250MG
3-(2-Methylpropyl)-1,2-oxazol-5-amine 95%
3-(2-Methylpropyl)-1,2-oxazol-5-amine
Formula: C₇H₁₂N₂O
MW: 140.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10692336
CAS Number: 1057064-36-4