(R)-1-(2,4-Dichlorophenyl)ethanamine 95%
Supplier: Apollo Scientific
OR944487-1G OR944487-250MG
APOSOR944487-1GEA 236 EUR
APOSOR944487-1G APOSOR944487-250MG
(R)-1-(2,4-Dichlorophenyl)ethanamine 95%
(R)-1-(2,4-Dichlorophenyl)ethanamine
Formula: C₈H₉Cl₂N
MW: 190.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06761788
CAS Number: 133773-29-2