3-[(Diethylamino)methyl]benzonitrile 98%
Supplier: Apollo Scientific
OR946392-1G OR946392-5G
APOSOR946392-1GEA 219 EUR
APOSOR946392-1G APOSOR946392-5G
3-[(Diethylamino)methyl]benzonitrile 98%
3-[(Diethylamino)methyl]benzonitrile
Formula: C₁₂H₁₆N₂
MW: 188.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09931537
CAS Number: 1016701-33-9