6-Methylisoindolin-1-one 95%

Supplier: Apollo Scientific

OR946562-1G OR946562-5G
APOSOR946562-1GEA 270 EUR
APOSOR946562-1G APOSOR946562-5G
6-Methylisoindolin-1-one 95%
6-Methylisoindolin-1-one
Formula: C₉H₉NO
Storage Temperature: Ambient
MDL Number: MFCD09056840
CAS Number: 58083-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR