5-Iodo-2-methyl-2,3-dihydropyridazin-3-one
Supplier: Apollo Scientific
OR949148-1G OR949148-500MG OR949148-5G
APOSOR949148-1GEA 655 EUR
APOSOR949148-1G APOSOR949148-500MG APOSOR949148-5G
5-Iodo-2-methyl-2,3-dihydropyridazin-3-one
5-Iodo-2-methyl-2,3-dihydropyridazin-3-one
Formula: C₅H₅IN₂O
MW: 236.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22052654
CAS Number: 153239-91-9