5-Chloro-2-(1,3-dioxolan-2-yl)-1,3-thiazole 95%
Supplier: Apollo Scientific
OR949387-1G OR949387-250MG
APOSOR949387-1GEA 806 EUR
APOSOR949387-1G APOSOR949387-250MG
5-Chloro-2-(1,3-dioxolan-2-yl)-1,3-thiazole 95%
5-Chloro-2-(1,3-dioxolan-2-yl)-1,3-thiazole
Formula: C₆H₆ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD23106372
CAS Number: 1080061-38-6