N-Formyl-4-iodo-L-phenylalanine
Supplier: Apollo Scientific
Warning
Synonyms: (2S)-2-(Formylamino)-3-(4-iodophenyl)propanoic acid, (2S)-2-(Formylamino)-3-(4-iodophenyl)propionic acid, N-[(1S)-1-Carboxy-2-(4-iodophenyl)ethyl]formamide
OR9507-5G OR9507-100G OR9507-25G
APOSOR9507-5GEA 29 EUR
APOSOR9507-5G APOSOR9507-100G APOSOR9507-25G
N-Formyl-4-iodo-L-phenylalanine
N-Formyl-4-iodo-L-phenylalanine
Formula: C₁₀H₁₀INO₃
MW: 319.096 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659491
CAS Number: 52721-77-4