3-(1H-Pyrazol-1-yl)phenol 95%
Supplier: Apollo Scientific
OR954194-1G OR954194-250MG
APOSOR954194-1GEA 647 EUR
APOSOR954194-1G APOSOR954194-250MG
3-(1H-Pyrazol-1-yl)phenol 95%
3-(1H-Pyrazol-1-yl)phenol
Formula: C₉H₈N₂O
MW: 160.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19686219
CAS Number: 904315-14-6