5-Ethoxy-N-methyl-2-nitroaniline 97%
Supplier: Apollo Scientific
OR954374-10G OR954374-1G OR954374-5G
APOSOR954374-10GEA 754 EUR
APOSOR954374-10G APOSOR954374-1G APOSOR954374-5G
5-Ethoxy-N-methyl-2-nitroaniline 97%
5-Ethoxy-N-methyl-2-nitroaniline
Formula: C₉H₁₂N₂O₃
MW: 196.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24448839
CAS Number: 1437794-71-2