4-[2-(Trimethylsilyl)ethynyl]morpholine 95%
Supplier: Apollo Scientific
Synonyms: 4-((Trimethylsilyl)ethynyl)morpholine
OR956569-1G OR956569-5G
APOSOR956569-1GEA 40.6 EUR
APOSOR956569-1G APOSOR956569-5G
4-[2-(Trimethylsilyl)ethynyl]morpholine 95%
4-[2-(Trimethylsilyl)ethynyl]morpholine
Formula: C₉H₁₇NOSi
MW: 183.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865184
CAS Number: 64024-63-1