4-Hydroxy-N-isopropylbenzamide 95%
Supplier: Apollo Scientific
OR957668-1G OR957668-5G
APOSOR957668-1GEA 270 EUR
APOSOR957668-1G APOSOR957668-5G
4-Hydroxy-N-isopropylbenzamide 95%
4-Hydroxy-N-isopropylbenzamide
Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00563307
CAS Number: 83191-67-7