N,N-Diethyl-3-nitroaniline 95%
Supplier: Apollo Scientific
Synonyms: N,N-Diethyl-m-nitroaniline
OR958268-1G OR958268-250MG
APOSOR958268-1GEA 179 EUR
APOSOR958268-1G APOSOR958268-250MG
N,N-Diethyl-3-nitroaniline 95%
N,N-Diethyl-3-nitroaniline
Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024459
CAS Number: 2216-16-2