4-(4-Nitrophenyl)but-3-en-2-one 95%
Supplier: Apollo Scientific
OR958451-1G OR958451-250MG
APOSOR958451-1GEA 178 EUR
APOSOR958451-1G APOSOR958451-250MG
4-(4-Nitrophenyl)but-3-en-2-one 95%
4-(4-Nitrophenyl)but-3-en-2-one
Formula: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00661313
CAS Number: 3490-37-7