1-Phenyl-1H-pyrazole-4-carbonitrile 95%

Supplier: Apollo Scientific

OR958630-1G OR958630-250MG
APOSOR958630-1GEA 0 EUR
APOSOR958630-1G APOSOR958630-250MG
1-Phenyl-1H-pyrazole-4-carbonitrile 95%
1-Phenyl-1H-pyrazole-4-carbonitrile
Formula: C₁₀H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD08691162
CAS Number: 709-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR