6-Methoxyimidazo[1,2-a]pyridine 98%
Supplier: Apollo Scientific
OR959480-1G OR959480-250MG
APOSOR959480-1GEA 806 EUR
APOSOR959480-1G APOSOR959480-250MG
6-Methoxyimidazo[1,2-a]pyridine 98%
6-Methoxyimidazo[1,2-a]pyridine
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11877992
CAS Number: 955376-51-9