Methyl 1-cyclopentylazetidine-3-carboxylate 98%
Supplier: Apollo Scientific
OR959920-1G
OR959920-5G
APOSOR959920-1GEA
328
EUR
APOSOR959920-1G
APOSOR959920-5G
Methyl 1-cyclopentylazetidine-3-carboxylate 98%
Methyl 1-cyclopentylazetidine-3-carboxylate
Formula:
C₁₀H₁₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD21333056 CAS Number: 1355247-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...